Please login to your online account to display your discounted pricing

anti-Trimethyl Histone H3 (Lys4), Clone: CMA304, EMD Millipore™


Manufacturer: emd millipore corporation  051339

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. 05-133-9MI

  • / Each
 

Description & Specifications

Specifications

Immunogen Synthetic peptide corresponding to amino acids 1-12 of human Histone H3, trimethylated on Lys4, conjugated to KLH.
Isotype κ
Antigen Trimethyl Histone H3 (Lys4)
Format Purified
Host Species Mouse
Applications Dot Blot
Applications ELISA
Applications Immunocytochemistry
Applications Immunoprecipitation
Applications Western Blotting
Purification Method Protein G purfied
Storage Requirements 2-8°C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles.
Monoclonal or Polyclonal Monoclonal
Quantity 100μg
Clone CMA304
Conjugate Unlabeled
Cross Reactivity Human
Cross Reactivity Vertebrate
Formulation Purified mouse monoclonal IgG1κ in PBS with 0.05% sodium azide.
Gene Accession No. NM_003493
Gene Symbols H3.4, H3/g , H3/t , H3FT , H3T, H3t , MGC126886, MGC126888 , OTTHUMP00000037945
Primary or Secondary Primary