Please login to your online account to display your discounted pricing

anti-Trimethyl Histone H3 (Lys4), Clone: CMA304, EMD Millipore™


Manufacturer: emd millipore corporation  051339S

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. 05-133-9SMI

  • / Each
 

Description & Specifications

Specifications

Immunogen Synthetic peptide corresponding to amino acids 1-12 of human Histone H3, trimethylated on Lys4, conjugated to KLH.
Isotype κ
Gene Symbols H3.4, H3/g , H3/t , H3FT , H3T, H3t , MGC126886, MGC126888 , OTTHUMP00000037945
Applications Dot Blot
Applications ELISA
Applications Immunocytochemistry
Applications Immunoprecipitation
Applications Western Blotting
Clone CMA304
Antigen Trimethyl Histone H3 (Lys4)
Format Purified
Formulation Purified mouse monoclonal IgG1κ in PBS with 0.05% sodium azide.
Gene Accession No. NM_003493
Host Species Mouse
Purification Method Protein G purfied
Quantity 10μg
Storage Requirements 2-8°C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles.
Primary or Secondary Primary
Conjugate Unlabeled
Cross Reactivity Human
Cross Reactivity Vertebrate
Monoclonal or Polyclonal Monoclonal