missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Trimethyl Histone H3 (Lys36) Rabbit anti-Human, Mouse, Polyclonal, MilliporeSigma™

Rabbit Polyclonal Antibody

$118.34 - $474.33

Specifications

Host Species Rabbit
Immunogen Linear peptide corresponding to human Histone H3 trimethylated at Lys36.
Antigen trimethyl Histone H3 (Lys36)
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
ABE305SMI View Documents MilliporeSigma
ABE305S
10μL Each for $118.34
 
ABE305MI View Documents MilliporeSigma
ABE305
100μL Each for $474.33
 
Description

Description

Use Anti-trimethyl Histone H3 (Lys36) Antibody (Rabbit Polyclonal Antibody) validated in western blot, DB, immunocytochemistry, Chimmunoprecipitation to detect trimethyl Histone H3 (Lys36) also known as H3K36me3, Histone H3 (tri methyl K36).
Specifications

Specifications

Rabbit
trimethyl Histone H3 (Lys36)
Affinity Purified
Chromatin Immunoprecipitation,Dot Blot,Immunocytochemistry,Western Blot
Purified rabbit polyclonal in buffer containing 0.1M Tris-Glycine (pH 7.4) and 150mM NaCl with 0.05% sodium azide.
Linear peptide corresponding to human Histone H3 trimethylated at Lys36.
Polyclonal
Primary
NP_003520

For Research Use Only