missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Trimethyl Histone H3 (Lys36) Rabbit anti-Human, Mouse, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Supplier: MilliporeSigma ABE305
Description
Use Anti-trimethyl Histone H3 (Lys36) Antibody (Rabbit Polyclonal Antibody) validated in western blot, DB, immunocytochemistry, Chimmunoprecipitation to detect trimethyl Histone H3 (Lys36) also known as H3K36me3, Histone H3 (tri methyl K36).Specifications
trimethyl Histone H3 (Lys36) | |
Polyclonal | |
Purified rabbit polyclonal in buffer containing 0.1M Tris-Glycine (pH 7.4) and 150mM NaCl with 0.05% sodium azide. | |
Rabbit | |
Affinity Purified | |
Primary | |
Human, Mouse |
ChIP Assay, Dot Blot, Immunocytochemistry, Western Blot | |
Unconjugated | |
HIST1H3A; H3FA; HIST1H3B; H3FL; HIST1H3C; H3FC; HIST1H3D; H3FB; HIST1H3E; H3FD; HIST1H3F; H3FI; HIST1H3G; H3FH; HIST1H3H; H3FK; HIST1H3I; H3FF; HIST1H3J; H3FJ | |
Linear peptide corresponding to human Histone H3 trimethylated at Lys36. | |
100 μL | |
NP_003520 | |
-20°C in undiluted aliquots for up to 12 months, Avoid Freeze/Thaw Cycles |
For Research Use Only