Please login to your online account to display your discounted pricing

anti-trimethyl Histone H3 (Lys36) Ab 1, Polyclonal, EMD Millipore™


Manufacturer: emd millipore corporation  07549S

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. 07-549-SMI

  • / Each
 

Description & Specifications

Specifications

Antigen trimethyl Histone H3 (Lys36)
Format Serum
Formulation Rabbit polyclonal antiserum IgG in buffer containing 0.05% sodium azide and 30% glycerol. Liquid at −20°C.
Gene Symbols H3F3A; MGC87783; H3.3A; MGC87782; H3F3; H3.3B; H3F3B
Host Species Rabbit
Applications Dot Blot
Applications Western Blotting
Immunogen KLH-conjugated
Isotype IgG
Purification Method Unpurified
Quantity 10μL
Storage Requirements −20°C in undiluted aliquots for 12 months. Avoid repeated freeze/thaw cycles.
Primary or Secondary Prrimary
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) NP_003484
Conjugate Unlabeled
Cross Reactivity Human