Please login to your online account to display your discounted pricing

anti-trimethyl Histone H3 (Lys27), Clone: 18E9.1, EMD Millipore™


Manufacturer: emd millipore corporation  051951S

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. 05-195-1SMI

  • / Each
 

Description & Specifications

Specifications

Isotype κ
Applications Immunocytochemistry
Applications Western Blotting
Primary or Secondary Primary
Antigen trimethyl Histone H3 (Lys27)
Monoclonal or Polyclonal Monoclonal
Purification Method Protein G Purified
Conjugate Unlabeled
Storage Requirements 2-8°C for one year
Clone 18E9.1
Format Purified
Quantity 10μL
Formulation Purified mouse monoclonal IgG2bκ in buffer containing 0.1MTris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Host Species Mouse
Gene Symbols HIST3H3, H3FT, H3/t, H3t, H3/g, H3.4, H3T
Cross Reactivity Human
Cross Reactivity Mouse
Gene Accession No. NP_002098
Immunogen Linear peptide corresponding to human Histone H3 trimethylated at Lys27.