Trimethyl Histone H3 (Lys27) Rabbit anti-Human, Mouse, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Manufacturer:  MilliporeSigma ABE44
 View more versions of this product
Catalog No. ABE44MI
Description
Description
Anti-trimethyl Histone H3 (Lys27) Antibody is a rabbit polyclonal antibody for detection of Histone H3 trimethylated on lysine 27 (H3K73me). This purified Ab is specificity verified by dot blot (DB) and validated in western blot, immunocytochemistry, PIA. Small pack size is ideal for screening applications.
Specifications
Specifications
trimethyl Histone H3 (Lys27)
Unconjugated
Affinity Purified
Rabbit
Affinity Purfied
2-8°C for 12 months
Polyclonal
Human,Murine
Dot Blot,Immunocytochemistry,Inhibition Assays,Western Blot
Purified rabbit polyclonal in buffer containing 0.1M Tris-Glycine (pH 7.4, 150mM NaCl) with 0.05% sodium azide.
HIST1H3A; H3FA; HIST1H3B; H3FL; HIST1H3C; H3FC; HIST1H3D; H3FB; HIST1H3E; H3FD; HIST1H3F; H3FI; HIST1H3G; H3FH; HIST1H3H; H3FK; HIST1H3I; H3FF; HIST1H3J; H3FJ; HIST3H3; H3FT; H3T; MGC126886
Branch peptide corresponding to human Histone H3 methylated at Lys27.
100μg
Primary
NP_003484.1