missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

anti-TRIF/TICAM1, Polyclonal, Novus Biologicals™

$139.00 - $362.00

Specifications

Host Species Rabbit
Immunogen This antibody was developed against a synthetic peptide corresponding to amino acids 143-158 of mouse TRIF (NP_778154).
Antigen TRIF/TICAM1
Monoclonal or Polyclonal Polyclonal
Format Antisera
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NB12013810T Novus Biologicals Canada
NB12013810SS
0.05mL Each for $139.00
NB12013810 Novus Biologicals Canada
NB12013810
0.2mL Each for $362.00
Specifications Description & Specifications

Specifications

Rabbit
TRIF/TICAM1
Antisera
RUO
The will cross-react with human TRIF, but additional bands may be seen. Cross reacts in Mouse. Human reactivity reported in the scientific literature (PMID: 23777661). Rat reactivity reported in scientific literature (PMID: 23447644).
PBS containing 0.05% BSA
This antibody was developed against a synthetic peptide corresponding to amino acids 143-158 of mouse TRIF (NP_778154).
Polyclonal
Primary
Immunocytochemistry,Immunofluorescence,Simple Western,Western Blot
148022