Please login to your online account to display your discounted pricing

anti-TRIB3, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB10056398SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB10056398T

  • / Each
 

Description & Specifications

Specifications

Test Specificity This detects a protein of approximate mol. wt. of 40 kDa in human liver lysate. Cross reacts with Human. Predicted to cross react with New World Monkey and Canine.
Concentration 0.5mg/mL
Gene Accession No. Q96RU7
Cross Reactivity Human
Applications Western Blotting
Regulatory Status RUO
Primary or Secondary Primary
Formulation PBS containing 0.05% BSA
Monoclonal or Polyclonal Polyclonal
Conjugate Unlabeled
Gene ID (Entrez) 57761
Immunogen A synthetic peptide corresponding to amino acid residues 315-332 of human TRIB3. Genbank accession number: NM_021158.
Antigen TRIB3
Host Species Rabbit
Quantity 0.025mg
Format Purified