Please login to your online account to display your discounted pricing

anti-TRIB3, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB10056398

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB10056398

  • / Each
 

Description & Specifications

Specifications

Concentration 0.5mg/mL
Formulation PBS containing 0.05% BSA
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 57761
Conjugate Unlabeled
Format Purified
Quantity 0.1mg
Antigen TRIB3
Applications Western Blotting
Test Specificity This detects a protein of approximate mol. wt. of 40 kDa in human liver lysate. Cross reacts with Human. Predicted to cross react with New World Monkey and Canine.
Primary or Secondary Primary
Regulatory Status RUO
Gene Accession No. Q96RU7
Immunogen A synthetic peptide corresponding to amino acid residues 315-332 of human TRIB3. Genbank accession number: NM_021158.
Cross Reactivity Human
Host Species Rabbit