missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

anti-TRIB3, Polyclonal, Novus Biologicals™

$139.00 - $255.80

Specifications

Host Species Rabbit
Immunogen A synthetic peptide corresponding to amino acid residues 315-332 of human TRIB3. Genbank accession number: NM_021158.
Antigen TRIB3
Monoclonal or Polyclonal Polyclonal
Format Purified
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NB10056398 Novus Biologicals Canada
NB10056398
0.1mg Each for $255.80
NB10056398T Novus Biologicals Canada
NB10056398SS
0.025mg Each for $139.00
Specifications Description & Specifications

Specifications

Rabbit
TRIB3
Purified
RUO
This detects a protein of approximate mol. wt. of 40 kDa in human liver lysate. Cross reacts with Human. Predicted to cross react with New World Monkey and Canine.
57761
0.5mg/mL
A synthetic peptide corresponding to amino acid residues 315-332 of human TRIB3. Genbank accession number: NM_021158.
Polyclonal
Primary
Western Blot
Q96RU7
PBS containing 0.05% BSA