missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

anti-TRHR, Polyclonal, Novus Biologicals™

$109.00 - $255.80

Specifications

Host Species Rabbit
Isotype IgG
Immunogen A portion of amino acids 350-400 of human TRH-R protein was used as the immunogen.
Antigen TRHR
Monoclonal or Polyclonal Polyclonal
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NBP224726 Novus Biologicals Canada
NBP224726
0.1mg Each for $255.80
NBP224726SS Novus Biologicals Canada
NBP224726SS
0.025mg Each for $109.00
Specifications Description & Specifications

Specifications

Rabbit
A portion of amino acids 350-400 of human TRH-R protein was used as the immunogen.
Polyclonal
Primary
Immunohistochemistry (Paraffin),Western Blot
7201
0.5mg/mL
IgG
TRHR
Purified
RUO
The amino acid sequence used as immunogen is 100% homologous in human, chimpanzee, rat, mouse and cow and 76% homologous in wild boar.
100μg in 200μL PBS containing 0.2% gelatin