Please login to your online account to display your discounted pricing

anti-TRHR, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP224726SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP224726SS

  • / Each
 

Description & Specifications

Specifications

Antigen TRHR
Applications Immunohistochemistry (Paraffin)
Applications Western Blot
Concentration 0.5mg/mL
Conjugate Unlabeled
Cross Reactivity Canine
Cross Reactivity Human
Cross Reactivity Primate
Format Purified
Formulation 100μg in 200μL PBS containing 0.2% gelatin
Host Species Rabbit
Immunogen A portion of amino acids 350-400 of human TRH-R protein was used as the immunogen.
Isotype IgG
Quantity 0.025mg
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 7201
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human, chimpanzee, rat, mouse and cow and 76% homologous in wild boar.
Target Species Canine
Target Species Human
Target Species Primate