Please login to your online account to display your discounted pricing

anti-TRF-1, Polyclonal, Novus Biologicals™

$139.00 - $255.80

Specifications

Host Species Rabbit
Immunogen Synthetic peptides corresponding to amino acids 321-335 and 406-420 of human TRF1 were used as immunogen.
Antigen TRF-1
Monoclonal or Polyclonal Polyclonal
Format Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB6001391

 
novus biologicals canada
NB6001391
Unlabeled 0.1mg Each for $255.80

NB6001391SS

 
novus biologicals canada
NB6001391SS
Unlabeled 0.025mg Each for $139.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen Synthetic peptides corresponding to amino acids 321-335 and 406-420 of human TRF1 were used as immunogen.
Antigen TRF-1
Monoclonal or Polyclonal Polyclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Western Blotting
Gene Accession No. P54274
Gene ID (Entrez) 7013
Formulation PBS with 0.05% BSA
Concentration 0.5mg/mL