Please login to your online account to display your discounted pricing

anti-TRF-1, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB6001391SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB6001391SS

  • / Each
 

Description & Specifications

Specifications

Gene ID (Entrez) 7013
Gene Accession No. P54274
Formulation PBS with 0.05% BSA
Format Purified
Immunogen Synthetic peptides corresponding to amino acids 321-335 and 406-420 of human TRF1 were used as immunogen.
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Concentration 0.5mg/mL
Conjugate Unlabeled
Regulatory Status RUO
Antigen TRF-1
Applications Western Blotting
Cross Reactivity Human
Host Species Rabbit
Quantity 0.025mg