Please login to your online account to display your discounted pricing

anti-TRF-1, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB6001391

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB6001391

  • / Each
 

Description & Specifications

Specifications

Quantity 0.1mg
Antigen TRF-1
Format Purified
Formulation PBS with 0.05% BSA
Gene Accession No. P54274
Host Species Rabbit
Applications Western Blotting
Immunogen Synthetic peptides corresponding to amino acids 321-335 and 406-420 of human TRF1 were used as immunogen.
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 7013
Concentration 0.5mg/mL
Conjugate Unlabeled
Cross Reactivity Human