Please login to your online account to display your discounted pricing

anti-TREX1, Polyclonal, Novus Biologicals™

$109.00 - $259.02

Specifications

Host Species Rabbit
Immunogen Partial recombinant protein made to an internal portion of human TREX1 protein (between residues 200-350) [NCBI NP_057465.1]
Antigen TREX1
Monoclonal or Polyclonal Polyclonal
Format Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP229617

 
novus biologicals canada
NBP229617
Unlabeled 0.1mg Each for $259.02

NBP229617SS

 
novus biologicals canada
NBP229617SS
Unlabeled 0.025mg Each for $109.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen Partial recombinant protein made to an internal portion of human TREX1 protein (between residues 200-350) [NCBI NP_057465.1]
Antigen TREX1
Monoclonal or Polyclonal Polyclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Western Blot
Gene Accession No. Q9NSU2
Gene ID (Entrez) 11277
Formulation PBS
Concentration 1.0mg/mL