Please login to your online account to display your discounted pricing

anti-TRBP, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB10056471

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB10056471

  • / Each
 

Description & Specifications

Specifications

Formulation PBS containing 0.05% BSA
Gene Accession No. Q15633
Primary or Secondary Primary
Concentration 0.5mg/mL
Conjugate Unlabeled
Cross Reactivity Bovine
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Primate
Cross Reactivity Rat
Cross Reactivity Zebrafish
Antigen TRBP
Format Affinity Purified
Host Species Rabbit
Applications Western Blotting
Immunogen A portion of amino acids 150-200 of human TRBP2 was used as the immunogen.
Quantity 0.1mg
Regulatory Status RUO
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 6895
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human (isoforms a and b), mouse, rat, bovine, chimp, and zebrafish. Human isoform a is a 366 amino acid protein, and human isoform b is a 345 amino acid protein. Cross reacts with Bovine, Chimpanzee, Human, Mouse, Rat and Zebrafish.