Please login to your online account to display your discounted pricing

anti-TRBP, Polyclonal, Novus Biologicals™

$139.00 - $259.02

Specifications

Host Species Rabbit
Isotype IgG
Immunogen A portion of amino acids 340-390 of human TRBP was used as the immunogen.
Antigen TRBP
Monoclonal or Polyclonal Polyclonal
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP224725

 
novus biologicals canada
NBP224725
Unlabeled 0.1mg Each for $259.02

NBP224725SS

 
novus biologicals canada
NBP224725SS
Unlabeled 0.025mg Each for $139.00
Description & Specifications

Specifications

Host Species Rabbit
Isotype IgG
Immunogen A portion of amino acids 340-390 of human TRBP was used as the immunogen.
Antigen TRBP
Monoclonal or Polyclonal Polyclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunohistochemistry (Paraffin)
Applications Western Blot
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human (isoforms CRA_a, CRA_f and CRA_d), rhesus monkey, chimpanzee and orangutan, 92% homologous in horse, cow and mouse and 85% homologous in dog and rat.
Gene Accession No. Q15633
Gene ID (Entrez) 6895
Formulation 100μg in 200μL PBS containing 0.2% gelatin
Concentration 0.5mg/mL