missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

anti-TRBP, Polyclonal, Novus Biologicals™

$139.00 - $255.80

Specifications

Host Species Rabbit
Immunogen A portion of amino acids 150-200 of human TRBP2 was used as the immunogen.
Antigen TRBP
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NB10056471 Novus Biologicals Canada
NB10056471
0.1mg Each for $255.80
NB10056471T Novus Biologicals Canada
NB10056471SS
0.025mg Each for $139.00
Specifications Description & Specifications

Specifications

Rabbit
TRBP
Affinity Purified
RUO
The amino acid sequence used as immunogen is 100% homologous in human (isoforms a and b), mouse, rat, bovine, chimp, and zebrafish. Human isoform a is a 366 amino acid protein, and human isoform b is a 345 amino acid protein. Cross reacts with Bovine, Chimpanzee, Human, Mouse, Rat and Zebrafish.
6895
0.5mg/mL
A portion of amino acids 150-200 of human TRBP2 was used as the immunogen.
Polyclonal
Primary
Western Blot
Q15633
PBS containing 0.05% BSA