missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

anti-TRAPPC9, Polyclonal, Novus Biologicals™

$139.00 - $255.80

Specifications

Host Species Rabbit
Immunogen Amino acids 282-299 (KKDFVGLDTDSRHYKKRC) of human NIBP was used as the immunogen.
Antigen TRAPPC9
Monoclonal or Polyclonal Polyclonal
Format Antisera
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NB10055735 Novus Biologicals Canada
NB10055735
0.1mL Each for $255.80
NB10055735T Novus Biologicals Canada
NB10055735SS
0.025mL Each for $139.00
Specifications Description & Specifications

Specifications

Rabbit
TRAPPC9
Antisera
RUO
The amino acid sequence used as immunogen is 100% homologous in human (isoform CRA_A and CRA_b), chimpanzee, monkey, mouse, opossum, rat and dog and 94% homologous in xenopus.
83696
Amino acids 282-299 (KKDFVGLDTDSRHYKKRC) of human NIBP was used as the immunogen.
Polyclonal
Primary
Immunocytochemistry,Immunofluorescence,Immunoprecipitation,Western Blot
Q96Q05
Rabbit sera with 50% glycerol