Please login to your online account to display your discounted pricing

anti-TRAPPC9, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB10055735SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB10055735T

  • / Each
 

Description & Specifications

Specifications

Test Specificity The amino acid sequence used as immunogen is 100% homologous in human (isoform CRA_A and CRA_b), chimpanzee, monkey, mouse, opossum, rat and dog and 94% homologous in xenopus.
Gene ID (Entrez) 83696
Primary or Secondary Primary
Cross Reactivity Bovine
Cross Reactivity Canine
Cross Reactivity Chicken
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Primate
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunoprecipitation
Applications Western Blotting
Regulatory Status RUO
Monoclonal or Polyclonal Polyclonal
Conjugate Unlabeled
Formulation Rabbit sera with 50% glycerol
Gene Accession No. Q96Q05
Immunogen Amino acids 282-299 (KKDFVGLDTDSRHYKKRC) of human NIBP was used as the immunogen.
Host Species Rabbit
Quantity 0.025mL
Antigen TRAPPC9
Format Antiserum