Please login to your online account to display your discounted pricing

anti-Transglutaminase 1/TGM1, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB1001844SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB1001844SS

  • / Each
 

Description & Specifications

Specifications

Antigen Transglutaminase 1/TGM1
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Western Blot
Conjugate Unlabeled
Cross Reactivity Human
Format Antisera
Gene Accession No. P22735
Host Species Rabbit
Immunogen A 15 residue synthetic peptide derived from a domainfrom human Transglutaminase 1 protein.
Quantity 0.02mL
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 7051
Test Specificity This reacts specifically with human Transglutaminase 1 . Not yet tested in other species.
Target Species Human