Please login to your online account to display your discounted pricing

anti-Transglutaminase 1/TGM1, Polyclonal, Novus Biologicals™

$110.17 - $329.00

Specifications

Host Species Rabbit
Immunogen A 15 residue synthetic peptide derived from a domainfrom human Transglutaminase 1 protein.
Antigen Transglutaminase 1/TGM1
Monoclonal or Polyclonal Polyclonal
Format Antisera
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB1001844

 
novus biologicals canada
NB1001844
Unlabeled 0.1mL Each for $329.00

NB1001844SS

 
novus biologicals canada
NB1001844SS
Unlabeled 0.02mL Each for $110.17
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A 15 residue synthetic peptide derived from a domainfrom human Transglutaminase 1 protein.
Antigen Transglutaminase 1/TGM1
Monoclonal or Polyclonal Polyclonal
Format Antisera
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Western Blot
Test Specificity This reacts specifically with human Transglutaminase 1 . Not yet tested in other species.
Gene Accession No. P22735
Gene ID (Entrez) 7051