Please login to your online account to display your discounted pricing
anti-Transferrin Receptor/CD71 Clone: H68.4, Thermo Scientific™ Novex™ DFS Item
Transferrin Receptor/CD71 Monoclonal antibody 13-6800 from Thermo Scientific™ Novex™ specifically detects Transferrin Receptor/CD71 in Chicken, Hamster, Mouse and Rat samples, and it is validated for WB, IF, IHC (P), FACS, IP, ELISA and IM
$454.00 - $982.00
Products
Catalog Number Mfr. No. Price Quantity    
136800
DFS Item
life technologies
LS136800
Each for $454.00
136890
DFS Item
life technologies
LS136890
Each for $982.00
Description & Specifications
CD71 (transferrin receptor) is a type II transmembrane glycoprotein expressed as homodimer in erythroid blood cell line and in activated leukocytes. Upon binding of holotransferrin (complex of transferrin and iron ions), CD71 is internalized by clathrin-mediated endocytosis. Acidification of endosomes by vesicular membrane proton pumps leads to dissociation of iron ions, whereas transferrin (apotransferrin) remains associated with CD71 and recycles to the cell surface, where it is released upon exposure to normal pH. CD71 is also involved in uptake of non-transferrin bound iron.
13-6800 detects Transferrin Receptor/CD71 in Chicken, Hamster, Mouse and Rat samples. 13-6800 has been successfully used in WB, IF, IHC (P), FACS, IP, ELISA and IM procedures. The immunogen for 13-6800 is recombinant human transferrin receptor.
Applications:
ELISA, FACS, Immunofluorescence, Immunohistochemistry (Paraffin), Immunomicroscopy, Immunoprecipitation, Western Blotting
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.