Please login to your online account to display your discounted pricing

anti-TRANCE/TNFSF11/RANK L, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB10056388

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB10056388

  • / Each
 

Description & Specifications

Specifications

Gene Accession No. NP_003692
Gene ID (Entrez) 8600
Concentration 0.5mg/mL
Primary or Secondary Primary
Formulation PBS containing 0.2% gelatin
Conjugate Unlabeled
Immunogen A recombinant protein containing amino acid residues 157-316 of mouse RANKL was used as immunogen.
Cross Reactivity Mouse
Quantity 0.1mg
Applications Western Blotting
Monoclonal or Polyclonal Polyclonal
Regulatory Status RUO
Antigen TRANCE/TNFSF11/RANK L
Format Purified
Host Species Rabbit