Please login to your online account to display your discounted pricing

anti-TRANCE/TNFSF11/RANK L, Polyclonal, Novus Biologicals™

$109.00 - $255.80

Specifications

Host Species Rabbit
Immunogen A recombinant protein containing amino acid residues 157-316 of mouse RANKL was used as immunogen.
Antigen TRANCE/TNFSF11/RANK L
Monoclonal or Polyclonal Polyclonal
Format Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB10056388T

 
novus biologicals canada
NB10056388SS
Unlabeled 0.025mg Each for $109.00

NB10056388

 
novus biologicals canada
NB10056388
Unlabeled 0.1mg Each for $255.80
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A recombinant protein containing amino acid residues 157-316 of mouse RANKL was used as immunogen.
Antigen TRANCE/TNFSF11/RANK L
Monoclonal or Polyclonal Polyclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Western Blotting
Gene Accession No. NP_003692
Gene ID (Entrez) 8600
Formulation PBS containing 0.2% gelatin
Concentration 0.5mg/mL