missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

anti-TRAIL/TNFSF10, Polyclonal, Novus Biologicals™

$142.00 - $142.00

Specifications

Host Species Rabbit
Immunogen This polyclonal antibody was raised against a peptide corresponding to amino acids 261-299 of human TRAIL.
Antigen TRAIL/TNFSF10
Monoclonal or Polyclonal Polyclonal
Format Purified
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NBP223644SS Novus Biologicals Canada
NBP223644SS
0.025mg Each for $142.00
Specifications Description & Specifications

Specifications

Rabbit
TRAIL/TNFSF10
Purified
RUO
8743
This polyclonal antibody was raised against a peptide corresponding to amino acids 261-299 of human TRAIL.
Polyclonal
Primary
Western Blot
0.1mg in 0.2mL PBS