Please login to your online account to display your discounted pricing

anti-TRAIL/TNFSF10, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP223644SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP223644SS

  • / Each
 

Description & Specifications

Specifications

Antigen TRAIL/TNFSF10
Format Purified
Formulation 0.1mg in 0.2mL PBS
Host Species Rabbit
Applications Western Blotting
Immunogen This polyclonal antibody was raised against a peptide corresponding to amino acids 261-299 of human TRAIL.
Quantity 0.025mg
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 8743
Conjugate Unlabeled
Cross Reactivity Human