missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

anti-TRAIL R3/TNFRSF10C, Polyclonal, Novus Biologicals™

$139.00 - $255.80

Specifications

Host Species Rabbit
Immunogen This antibody was developed against a synthetic peptide corresponding to amino acids 155-168 ([BLAST]CSSSVSNPRNWLFL[/BLAST]) of mouse DcTrailR1; GenBank no: NP_077252.2.
Antigen TRAIL R3/TNFRSF10C
Monoclonal or Polyclonal Polyclonal
Format Purified
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NB10056445 Novus Biologicals Canada
NB10056445
0.1mg Each for $255.80
NB10056445T Novus Biologicals Canada
NB10056445SS
0.025mg Each for $139.00
Specifications Description & Specifications

Specifications

Rabbit
TRAIL R3/TNFRSF10C
Purified
RUO
In mouse spleen, a 20 kDa band is observed. The amino acid sequence used is 92% homologous in DcTrailR2.
PBS containing 0.2% gelatin
This antibody was developed against a synthetic peptide corresponding to amino acids 155-168 ([BLAST]CSSSVSNPRNWLFL[/BLAST]) of mouse DcTrailR1; GenBank no: NP_077252.2.
Polyclonal
Primary
Western Blot
8794
0.5mg/mL