Please login to your online account to display your discounted pricing

anti-TRAIL R2/TNFRSF10B, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB10056618

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB10056618

  • / Each
 

Description & Specifications

Specifications

Antigen TRAIL R2/TNFRSF10B
Format Purified
Formulation PBS containing 0.05% BSA
Host Species Rabbit
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Immunogen 2). The same sequence is found in isoform 1 at amino acids 417-436.
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 8795
Concentration 0.5mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Quantity 0.1mg