Please login to your online account to display your discounted pricing

anti-TRAF7, Polyclonal, Novus Biologicals™

$139.00 - $274.59

Specifications

Host Species Rabbit
Isotype IgG
Immunogen Amino acids 334-356 of human TRAF7 protein were used as the immunogen for this antibody.
Antigen TRAF7
Monoclonal or Polyclonal Polyclonal
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB6001399

 
novus biologicals canada
NB6001399
Unlabeled 0.1mg Each for $274.59

NB6001399SS

 
novus biologicals canada
NB6001399SS
Unlabeled 0.025mg Each for $139.00
Description & Specifications

Specifications

Host Species Rabbit
Isotype IgG
Immunogen Amino acids 334-356 of human TRAF7 protein were used as the immunogen for this antibody.
Antigen TRAF7
Monoclonal or Polyclonal Polyclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunoprecipitation
Applications Western Blot
Test Specificity Please check J Biol Chem. 2011 Jul 1;286(26):22924-33. Epub 2011 Apr 25 for specific Images and Application data. The amino acid sequence used as immunogen is 100% identical in human (isoforms CRA_a and CRA_c), mouse, rat and horse.
Gene ID (Entrez) 84231
Formulation PBS containing 0.05% BSA
Concentration 0.5mg/mL