Please login to your online account to display your discounted pricing

anti-TRAF3IP2, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB10056740

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB10056740

  • / Each
 

Description & Specifications

Specifications

Gene Accession No. NP_679211
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Test Specificity An approx. 60 kDa band is observed. Cross reacts with Mouse and Human. Predicted to react with Rat. The amino acid sequence used as immunogen for this antibody is 100% homologous in human, 88% in rat, and 82% in mouse.
Concentration 0.5mg/mL
Antigen TRAF3IP2
Format Purified
Formulation PBS containing 0.05% BSA
Host Species Rabbit
Applications Immunohistochemistry (Paraffin)
Applications Simple Western
Applications Western Blotting
Immunogen This antibody was developed against a synthetic peptide (QDLPRPLRSREFPQFEP) corresponding to amino acids 225-241 of human TRAF3IP2.
Quantity 0.1mg
Regulatory Status RUO
Gene ID (Entrez) 10758
Conjugate Unlabeled
Cross Reactivity Human
Cross Reactivity Mouse