Please login to your online account to display your discounted pricing

anti-TRAF3IP1, Polyclonal, Novus Biologicals™

$139.00 - $255.80

Specifications

Host Species Rabbit
Immunogen A portion of amino acids 40-90 from human TRAF3IP1 was used as the immunogen for this antibody.NP_056465.2
Antigen TRAF3IP1
Monoclonal or Polyclonal Polyclonal
Format Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP219011

 
novus biologicals canada
NBP219011
Unlabeled 0.1mg Each for $255.80

NBP219011SS

 
novus biologicals canada
NBP219011SS
Unlabeled 0.025mg Each for $139.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A portion of amino acids 40-90 from human TRAF3IP1 was used as the immunogen for this antibody.NP_056465.2
Antigen TRAF3IP1
Monoclonal or Polyclonal Polyclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Western Blot
Test Specificity The TRAF3IP1 amino acid sequence used as immunogen for this antibody is 100% homologous in human, rat, pig, pole-cat, guinea pig, dog, horse, macaque, rhesus monkey, white-cheeked gibbon, chimpanzee, and orangutan, 94% in panda, cow, new world monkey, Chinese hamster, and mouse, 89% in Carolina anole, chicken, wild turkey, opossum, and platypus, 83% in zebrafish, zebra finch, and frog and 72% homologouus in cichlid fish.
Gene ID (Entrez) 26146
Formulation PBS containing 0.05% BSA