missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

anti-TRAF3IP1, Polyclonal, Novus Biologicals™

$139.00 - $255.80

Specifications

Host Species Rabbit
Immunogen A portion of amino acids 390-440 from human TRAF3IP1 was used as the immunogen for this antibody.
Antigen TRAF3IP1
Monoclonal or Polyclonal Polyclonal
Format Purified
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NBP214847 Novus Biologicals Canada
NBP214847
0.1mg Each for $255.80
NBP214847SS Novus Biologicals Canada
NBP214847SS
0.025mg Each for $139.00
Specifications Description & Specifications

Specifications

Rabbit
TRAF3IP1
Purified
RUO
The TRAF3IP1 amino acid sequence used as immunogen for this antibody is 100% homologous in human, and chimpanzee, 89% in orangutan, and white-cheeked gibbon, 74% in rhesus monkey, 68% in new world monkey, and 63% in panda, opossum, and rat.
26146
0.5mg/mL
A portion of amino acids 390-440 from human TRAF3IP1 was used as the immunogen for this antibody.
Polyclonal
Primary
Immunohistochemistry,Immunohistochemistry (Paraffin)
Q8TDR0
PBS containing 0.05% BSA