Please login to your online account to display your discounted pricing

anti-TRAF3IP1, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP219011SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP219011SS

  • / Each
 

Description & Specifications

Specifications

Primary or Secondary Primary
Formulation PBS containing 0.05% BSA
Gene ID (Entrez) 26146
Test Specificity The TRAF3IP1 amino acid sequence used as immunogen for this antibody is 100% homologous in human, rat, pig, pole-cat, guinea pig, dog, horse, macaque, rhesus monkey, white-cheeked gibbon, chimpanzee, and orangutan, 94% in panda, cow, new world monkey, Chinese hamster, and mouse, 89% in Carolina anole, chicken, wild turkey, opossum, and platypus, 83% in zebrafish, zebra finch, and frog and 72% homologouus in cichlid fish.
Conjugate Unlabeled
Format Purified
Immunogen A portion of amino acids 40-90 from human TRAF3IP1 was used as the immunogen for this antibody.NP_056465.2
Regulatory Status RUO
Quantity 0.025mg
Monoclonal or Polyclonal Polyclonal
Antigen TRAF3IP1
Applications Western Blotting
Host Species Rabbit
Cross Reactivity Human