Please login to your online account to display your discounted pricing

anti-TRAF-2, Polyclonal, Novus Biologicals™

$144.63 - $222.95

Specifications

Host Species Rabbit
Immunogen A synthetic peptide corresponding to amino acids 250-271(CSFLEAQASPGTLNQVGPELLQR) of mouse TRAF2 was used as immunogen
Antigen TRAF-2
Monoclonal or Polyclonal Polyclonal
Format Antisera
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB10056174

 
novus biologicals canada
NB10056174
Unlabeled 0.05mL Each for $222.95

NB10056174T

 
novus biologicals canada
NB10056174SS
Unlabeled 0.025mL Each for $144.63
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A synthetic peptide corresponding to amino acids 250-271(CSFLEAQASPGTLNQVGPELLQR) of mouse TRAF2 was used as immunogen
Antigen TRAF-2
Monoclonal or Polyclonal Polyclonal
Format Antisera
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blot
Test Specificity The gene encoding TRAF2 produces, by alternative splicing, 15 different transcripts, all with introns, putatively encoding 15 different protein isoforms. These isoforms are of varying amino acid lengths. Therefore the molecular weight observed by western blot may vary, depending on the isoform expressed. Users are encouraged to use the peptide immunogen sequence information provided in the antigen section to gain additional information about the potential TRAF2 isoforms detected by the antibody.
Gene ID (Entrez) 7186
Formulation PBS