Please login to your online account to display your discounted pricing

anti-TRAF-2, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB10056174

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB10056174

  • / Each
 

Description & Specifications

Specifications

Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Immunogen A synthetic peptide corresponding to amino acids 250-271(CSFLEAQASPGTLNQVGPELLQR) of mouse TRAF2 was used as immunogen
Primary or Secondary Primary
Test Specificity The gene encoding TRAF2 produces, by alternative splicing, 15 different transcripts, all with introns, putatively encoding 15 different protein isoforms. These isoforms are of varying amino acid lengths. Therefore the molecular weight observed by western blot may vary, depending on the isoform expressed. Users are encouraged to use the peptide immunogen sequence information provided in the antigen section to gain additional information about the potential TRAF2 isoforms detected by the antibody.
Conjugate Unlabeled
Antigen TRAF-2
Format Antiserum
Formulation PBS
Host Species Rabbit
Quantity 0.05mL
Regulatory Status RUO
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 7186
Cross Reactivity Mouse