missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

anti-Trace Amine Receptor 1, Polyclonal, Novus Biologicals™

$109.00 - $274.59

Specifications

Host Species Rabbit
Isotype IgG
Immunogen A portion of amino acids 225-250 of human TAAR1 was used as the immunogen
Antigen Trace Amine Receptor 1
Monoclonal or Polyclonal Polyclonal
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NBP224720SS Novus Biologicals Canada
NBP224720SS
0.025mg Each for $109.00
NBP224720 Novus Biologicals Canada
NBP224720
0.1mg Each for $274.59
Specifications Description & Specifications

Specifications

Rabbit
A portion of amino acids 225-250 of human TAAR1 was used as the immunogen
Polyclonal
Primary
Immunohistochemistry (Paraffin),Western Blot
Q96RJ0
100μg in 200μL PBS containing 0.05% BSA
IgG
Trace Amine Receptor 1
Purified
RUO
The amino acid sequence used as immunogen is 100% homologous in human and chimpanzee, 86% homologous in mouse and 80% homologous in dog.
134864
1mg/mL