Please login to your online account to display your discounted pricing

anti-TPX2, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB500179SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB500179SS

  • / Each
 

Description & Specifications

Specifications

Antigen TPX2
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunoprecipitation
Applications Simple Western
Applications Western Blot
Conjugate Unlabeled
Cross Reactivity Human
Cross Reactivity Mammalia
Cross Reactivity Mouse
Cross Reactivity Porcine
Cross Reactivity Rat
Format Antisera
Gene Accession No. Q9ULW0
Host Species Rabbit
Immunogen A recombinant segment of the C-terminal domain of human TPX2 [UniProt No. Q9ULW0]
Quantity 0.025mL
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 22974
Target Species Human
Target Species Mammalia
Target Species Murine
Target Species Porcine
Target Species Rat