Please login to your online account to display your discounted pricing

anti-TPX2, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB500179

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB500179

  • / Each
 

Description & Specifications

Specifications

Monoclonal or Polyclonal Polyclonal
Primary or Secondary Primary
Quantity 0.1mL
Cross Reactivity Human
Cross Reactivity Mammalia
Cross Reactivity Mouse
Cross Reactivity Porcine
Cross Reactivity Rat
Antigen TPX2
Immunogen A recombinant segment of the C-terminal domain of human TPX2 [UniProt No. Q9ULW0]
Regulatory Status RUO
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunoprecipitation
Applications Simple Western
Applications Western Blotting
Conjugate Unlabeled
Gene Accession No. Q9ULW0
Gene ID (Entrez) 22974
Host Species Rabbit
Format Antiserum