missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

anti-TPX2, Polyclonal, Novus Biologicals™

$139.00 - $329.00

Specifications

Host Species Rabbit
Immunogen A recombinant segment of the C-terminal domain of human TPX2 [UniProt No. Q9ULW0]
Antigen TPX2
Monoclonal or Polyclonal Polyclonal
Format Antisera
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NB500179 Novus Biologicals Canada
NB500179
0.1mL Each for $329.00
NB500179SS Novus Biologicals Canada
NB500179SS
0.025mL Each for $139.00
Specifications Description & Specifications

Specifications

Rabbit
TPX2
Antisera
RUO
Q9ULW0
A recombinant segment of the C-terminal domain of human TPX2 [UniProt No. Q9ULW0]
Polyclonal
Primary
Flow Cytometry,Immunocytochemistry,Immunofluorescence,Immunoprecipitation,Simple Western,Western Blot
22974