Please login to your online account to display your discounted pricing

anti-TP53INP2, Polyclonal, Novus Biologicals™

$109.00 - $309.05

Specifications

Host Species Rabbit
Immunogen A genomic peptide made to a C-terminal region of the human TP53INP2 protein (within residues 100-220). [Swiss-Prot Q8IXH6]
Antigen TP53INP2
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP171704SS

 
novus biologicals canada
NBP171704SS
Unlabeled 0.025mL Each for $109.00

NBP171704

 
novus biologicals canada
NBP171704
Unlabeled 0.1mL Each for $309.05
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A genomic peptide made to a C-terminal region of the human TP53INP2 protein (within residues 100-220). [Swiss-Prot Q8IXH6]
Antigen TP53INP2
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Test Specificity Human and mouse. Immunogen has 81% identity to rat.
Gene ID (Entrez) 58476
Formulation PBS, 30% glycerol
Concentration 0.6mg/mL