Please login to your online account to display your discounted pricing

anti-TNIP2, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB10056701SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB10056701T

  • / Each
 

Description & Specifications

Specifications

Gene ID (Entrez) 79155
Gene Accession No. Q8NFZ5
Conjugate Unlabeled
Concentration 0.5mg/mL
Formulation PBS containing 0.05% BSA
Quantity 0.025mg
Antigen TNIP2
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Immunogen This antibody was generated by immunizing rabbits with synthetic peptides of human ABIN-2 (NP_077285).
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Host Species Rabbit
Format Purified
Regulatory Status RUO
Applications Western Blotting