Please login to your online account to display your discounted pricing

anti-TNIP2, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB10056701

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB10056701

  • / Each
 

Description & Specifications

Specifications

Concentration 0.5mg/mL
Formulation PBS containing 0.05% BSA
Conjugate Unlabeled
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Antigen TNIP2
Host Species Rabbit
Regulatory Status RUO
Quantity 0.1mg
Immunogen This antibody was generated by immunizing rabbits with synthetic peptides of human ABIN-2 (NP_077285).
Gene ID (Entrez) 79155
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Gene Accession No. Q8NFZ5
Format Purified
Applications Western Blotting