Please login to your online account to display your discounted pricing

anti-TNFAIP2, Polyclonal, Novus Biologicals™

$109.00 - $255.80

Specifications

Host Species Rabbit
Immunogen A portion of amino acids 320-370 from human TNFAIP2 was used as the immunogen for this antibody.
Antigen TNFAIP2
Monoclonal or Polyclonal Polyclonal
Format Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP227085

 
novus biologicals canada
NBP227085
Unlabeled 0.1mg Each for $255.80

NBP227085SS

 
novus biologicals canada
NBP227085SS
Unlabeled 0.025mg Each for $109.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A portion of amino acids 320-370 from human TNFAIP2 was used as the immunogen for this antibody.
Antigen TNFAIP2
Monoclonal or Polyclonal Polyclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunohistochemistry (Paraffin)
Test Specificity The amino acid sequence used as immunogen for this antibody is 100% homologous in human, and chimpanzee, 94% in white-cheeked gibbon, and orangutan, 81% in new world monkey, rhesus monkey, 69% in dog, Chinese hamster, african bush elephant, and rat, 62% homologous in pole-cat, mouse, mole rat, guinea pig, and panda.
Gene Accession No. Q03169
Gene ID (Entrez) 7127
Formulation 0.1mg antibody in 0.2mL PBS containing 0.05% BSA.
Concentration 0.5mg/mL