Please login to your online account to display your discounted pricing

anti-TNFAIP2, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP227085SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP227085SS

  • / Each
 

Description & Specifications

Specifications

Gene Accession No. Q03169
Concentration 0.5mg/mL
Formulation 0.1mg antibody in 0.2mL PBS containing 0.05% BSA.
Primary or Secondary Primary
Conjugate Unlabeled
Monoclonal or Polyclonal Polyclonal
Regulatory Status RUO
Cross Reactivity Human
Cross Reactivity Primate
Host Species Rabbit
Applications Immunohistochemistry (Paraffin)
Test Specificity The amino acid sequence used as immunogen for this antibody is 100% homologous in human, and chimpanzee, 94% in white-cheeked gibbon, and orangutan, 81% in new world monkey, rhesus monkey, 69% in dog, Chinese hamster, african bush elephant, and rat, 62% homologous in pole-cat, mouse, mole rat, guinea pig, and panda.
Gene ID (Entrez) 7127
Quantity 0.025mg
Antigen TNFAIP2
Immunogen A portion of amino acids 320-370 from human TNFAIP2 was used as the immunogen for this antibody.
Format Purified