Please login to your online account to display your discounted pricing

anti-TNFAIP2, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP227085

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP227085

  • / Each
 

Description & Specifications

Specifications

Gene Accession No. Q03169
Concentration 0.5mg/mL
Antigen TNFAIP2
Format Purified
Formulation 0.1mg antibody in 0.2mL PBS containing 0.05% BSA.
Host Species Rabbit
Applications Immunohistochemistry (Paraffin)
Immunogen A portion of amino acids 320-370 from human TNFAIP2 was used as the immunogen for this antibody.
Quantity 0.1mg
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 7127
Test Specificity The amino acid sequence used as immunogen for this antibody is 100% homologous in human, and chimpanzee, 94% in white-cheeked gibbon, and orangutan, 81% in new world monkey, rhesus monkey, 69% in dog, Chinese hamster, african bush elephant, and rat, 62% homologous in pole-cat, mouse, mole rat, guinea pig, and panda.
Conjugate Unlabeled
Cross Reactivity Human
Cross Reactivity Primate