missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

anti-TNF-alpha, Polyclonal, Novus Biologicals™

$110.17 - $222.95

Specifications

Host Species Rabbit
Immunogen Recombinant full length protein produced in E. coli (Human).
Antigen TNF-alpha
Monoclonal or Polyclonal Polyclonal
Format Antisera
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NB600587SS Novus Biologicals Canada
NB600587SS
0.04mL Each for $110.17
NB600587 Novus Biologicals Canada
NB600587
0.2mL Each for $222.95
Specifications Description & Specifications

Specifications

Rabbit
TNF-alpha
Antisera
RUO
The antiserum is directed against mature 17,000 MW human TNFa and is useful in determining its presence in various assays. In general, this antibody also detects primate TNFa in the same formats using similar dilutions. The antibody does not recognize human TNG beta (lymphotoxin). This antiserum will recognize the cell-bound precursor of TNFa as a 26,000 protein in immunoblots, particularly in denatured samples. This antibody is also useful for neutralization of human and primate TNFa activity in bioassays. It does not neutralize the biological activity of lymphotoxin. For neutralization, it is recommended to incubate the sample with a 1:200 dilution of the antibody for at least 4 hours before being tested. A control of similarly diluted normal rabbit IgG is recommended.
Whole antisera; total protein concentration: 85mg/mL by Refractometry.
Recombinant full length protein produced in E. coli (Human).
Polyclonal
Primary
ELISA,Immunohistochemistry,Immunohistochemistry (Paraffin),Western Blot
7124