Please login to your online account to display your discounted pricing

anti-TNF-alpha, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP119532SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP119532SS

  • / Each
 

Description & Specifications

Specifications

Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 7124
Gene Accession No. P01375
Regulatory Status RUO
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Quantity 0.025mg
Immunogen A synthetic peptide made to an internal portion of the human TNF alpha protein (between residues 100-200) [UniProt P01375]
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Test Specificity Human, Mouse and Rat. Immunogen has 100% homology with porcine and feline, and 90% homology to mink, canine and monkey.
Primary or Secondary Primary
Conjugate Unlabeled
Formulation PBS
Concentration 1.0mg/mL
Host Species Rabbit
Format Affinity Purified
Antigen TNF-alpha